Attune® 96-well Plate

Catalog number: 4477131

Applied Biosystems™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This 96-well plate is used on the Attune® Auto Sampler. It is a non-treated polystyrene plate with U-bottom wells and a low-evaporation lid.

Features include:

• Bacteriological-grade polystyrene that is non-treated for a more hydrophobic growth surface and reduced cell attachment
• Alphanumeric well identification
• Condensation rings
• Lid for reduced evaporation and contamination
• Individually packaged in convenient peel-open medical-style packaging (recyclable PET trays)
• Sterilized by gamma irradiation, nonpyrogenic
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Format: 96-well plate
Product Line: Attune®
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Plate, Plate Lid or Cover

Contents & storage

Store at room temperature

Documents

Manuals & protocols