Cadmium sulfate, ACS reagent, anhydrous
Cadmium sulfate, ACS reagent, anhydrous
Cadmium sulfate, ACS reagent, anhydrous
Thermo Scientific Chemicals

Cadmium sulfate, ACS reagent, anhydrous

CAS: 10124-36-4 | CdO4S | 208.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447755000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10124-36-4
IUPAC Namecadmium(2+) sulfate
Molecular FormulaCdO4S
InChI KeyQCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES[Cd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Insoluble matter=<0.005 %
Loss on drying=<1.0 % (1 g, 150°C, 4 h)
Chloride (Cl)=<0.001 %
Nitrate and Nitrite (as NO3)=<0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only