The Ion TargetSeq Exome workflow offers standard and accelerated protocols to generate a library in 4 or 2 days, respectively. Suggested timelines for generating TargetSeq Exome Libraries are shown below;
Standard protocol timeline;
Day 1: Prepare library, amplify and purify, quanitfy, set up and start hybridization (10 hrs)
Days 2-4: Hybridization at 47 degrees C (64-72 hrs)
Day 4: Wash and recover beads with enriched DNA, amplify and purify, quantify the enriched library (5 hrs)
Accelerated protocol timeline;
Day 1: Prepare library, amplify and purify, quanitfy, setup and start hybridization (10 hrs)
Night 1: Hybridization at 47 degrees C (16-18 hrs)
Day 2: Wash and recover beads with enriched DNA, amplify and purify, quantify the enriched library (5 hrs)
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.