5500 W Conversion Primers Kit

Catalog number: 4478020

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 W Conversion Primers Kit is used to prepare libraries for sequencing on the 5500 W Series genetic analyzers.

The 5500 W Conversion Primers Kit contains primers to amplify the library and enable the library to be amplified on the 5500 W FlowChip for sequencing. The kit also includes an enzyme to modify the library prior to deposition of the library into the 5500 W FlowChip.

The 5500 W Conversion Primers Kit is compatible with all 5500 library construction kits. This kit contains sufficient reagents to prepare 12 libraries.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Library Type: Fragment Library, Mate-Paired Library, Targeted Sequencing Library, cDNA Library
Workflow Step: Library Generation
Sequencing Type: Epigenetic Sequencing, Genome & DNA Sequencing, Small RNA Sequencing, Transcriptome Sequencing
Product Line: SOLiD™
Product Size: 12 reactions
Shipping Condition: Wet Ice

Contents & storage

Store at -15 to -25 °C

Documents

Manuals & protocols