Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Thermo Scientific Chemicals

Sodium benzoate, 99%, for biochemistry

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 447802500
Price (USD)
63.65
Online Exclusive
70.30
Save 6.65 (9%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.30
Save 6.65 (9%)
Each
Chemical Identifiers
CAS532-32-1
IUPAC Namesodium benzoate
Molecular FormulaC7H5NaO2
InChI KeyWXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration with HClO4>=98.5 %
Loss on drying=<1 %
Solubility(1 M in water) Clear colorless to light yellow
Mercury (Hg)=<0.0001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only