Sodium sulfate decahydrate, 99%, for biochemistry
Sodium sulfate decahydrate, 99%, for biochemistry
Sodium sulfate decahydrate, 99%, for biochemistry
Thermo Scientific Chemicals

Sodium sulfate decahydrate, 99%, for biochemistry

CAS: 7727-73-3 | Na2O4S·10H2O | 322.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 447930025
Price (USD)
241.65
Special offer
Online exclusive
Ends: 28-Dec-2026
284.00
Save 42.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
241.65
Special offer
Online exclusive
Ends: 28-Dec-2026
284.00
Save 42.35
Each
Chemical Identifiers
CAS7727-73-3
IUPAC Namedisodium decahydrate sulfate
Molecular FormulaH20Na2O14S
InChI KeyRSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<0.001 %
UV(1 M in water)
UVat 260 nm A: =<0.025
UVat 280 nm A: =<0.021
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only