4N6FLOQSwabs™ for Crime Scene (regular tip, tube)

Catalog number: 4479427

 Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

At crime scenes, it is essential that the sample collection device prevents microbial contamination while preserving nucleic acid integrity. The unique 4N6FLOQSwabs™ for Crime Scene product line is specifically designed for sample collection of sweat, semen, blood stains, skin, and environmental trace evidence found at the crime scene. 4N6FLOQSwabs™ for Crime Scene feature short Nylon® fibers that are arranged in a perpendicular fashion to create a highly absorbent thin layer with an open structure to facilitate sample elution. 4N6FLOQSwabs™ for Crime Scene (Regular Tip, Tube) have regular-size tips. Each swab is packaged in a plastic tube to guarantee swab integrity and avoid risks of sample contamination during transport.

Key Features:

• Certified free of DNase, RNase and amplifiable human DNA
• Rapid absorption capability and superior sample release
• Antimicrobial treatment to prevent microbial growth and improve sample preservation
• No need for sample drying
• Break Point (from tip end): 20 mm

4N6FLOQSwabs™ from Crime Scene are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
For Forensic or Paternity Use Only.

Specifications

Format: Tube(s)
Product Line: 4N6FLOQSwabs™
Product Size: Case of 100
Shipping Condition: Approved for shipment at Room Temperature or on Dry Ice
Type: Sample Collection Accessory

Contents & storage

100 Regular head tip FLOQSwabs™, in plastic tube, with Antimicrobial Action
Store at RT

Documents

Manuals & protocols