4N6FLOQSwabs™, regular tip, tube
Applied Biosystems™
4N6FLOQSwabs™, regular tip, tube
4N6FLOQSwabs, regular tip, are comprised of a solid, molded plastic applicator shaft with a regular-sized tip. The tip of theMás información
Have Questions?
Número de catálogoCantidad
4479433100 hisopos
Número de catálogo 4479433
Precio (MXN)
3,299.32
Each
Cantidad:
100 hisopos
Precio (MXN)
3,299.32
Each
4N6FLOQSwabs, regular tip, are comprised of a solid, molded plastic applicator shaft with a regular-sized tip. The tip of the applicator is coated with short Nylon fibers that are arranged in a perpendicular fashion. This perpendicular arrangement results from a process called flocking, where the fibers are sprayed onto the tip of the swab while it is held in an electrostatic field. This process creates a highly absorbent, thin layer with an open structure. The swab is supplied in a dry tube that helps prevent sample contamination during transport and is suitable for reference and evidence collection applications.
Key features:
• Certified free of DNase, RNase, and amplifiable human DNA
• Rapid absorption capability and superior sample release
• Regular-size tip for buccal cell collection
• Break point (from tip end): 20 mm
Unlike traditional fiber wound swabs, 4N6FLOQSwabs have no internal absorbent core to disperse and entrap the specimen—the sample stays close to the surface for faster and more efficient elution. The perpendicular Nylon fibers act like a soft brush that facilitates improved collection of cellular material. Capillary action between the fiber strands facilitates strong hydraulic uptake of liquid sample, and the sample stays close to the surface allowing easy elution.
4N6FLOQSwabs are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
Especificaciones
Línea de productos4N6FLOQSwabs
Cantidad100 hisopos
Condiciones de envíoTemperatura ambiente
Applicator MaterialNylon
Material de la puntaNylon Fiber
Tipo de puntaRegular
Unit SizeEach
Contenido y almacenamiento
Store at room temperature.