Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
Thermo Scientific Chemicals

Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous

CAS: 7558-80-7 ∣ H2NaO4P ∣ 119.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 447982500
Price (USD)
60.65
Online exclusive
66.90
Save 6.25
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
60.65
Online exclusive
66.90
Save 6.25
Each
Chemical Identifiers
CAS7558-80-7
IUPAC Namesodium dihydrogen phosphate
Molecular FormulaH2NaO4P
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.2 % (in water)
pH4.0 to 6.0 (1 M in water)
UV(1 M in water)
UVat 260 nm A: =<0.2
Iron (Fe)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only