Potassium thiocyanate, 99%, for biochemistry
Potassium thiocyanate, 99%, for biochemistry
Potassium thiocyanate, 99%, for biochemistry
Thermo Scientific Chemicals

Potassium thiocyanate, 99%, for biochemistry

CAS: 333-20-0 | CKNS | 97.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 448000010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.005 %
Ammonium (NH4)=<0.003 %
Appearance (Color)White
Titration Argentometric>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only