Citric acid, diammonium salt, 99+%, for biochemistry
Citric acid, diammonium salt, 99+%, for biochemistry
Citric acid, diammonium salt, 99+%, for biochemistry
Thermo Scientific Chemicals

Citric acid, diammonium salt, 99+%, for biochemistry

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 448021000
Price (EUR)
42,50
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
42,50
Each
Chemical Identifiers
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
SolubilityClear colorless
Heavy metals (as Pb)=<5 ppm
UV(1 M in water)
UVat 280 nm A: =<0.1
Chloride (Cl)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only