Ion AmpliSeq™ Library Kit 2.0
Ion Torrent™
Ion AmpliSeq™ Library Kit 2.0
The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수
448044196 Reactions
44753458 Reactions
4480442384 Reactions
카탈로그 번호 4480441
제품 가격(KRW)
-
반응 수:
96 Reactions
The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.
The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ ready-to-use panels and custom primer pools, for sequencing on the Ion Personal Genome Machine™ (PGM™) System. This new 'LV' version of the kit is configured for large reagent volumes, making it ideal for automated DNA library preparation protocols; it can also be used with manual protocols. This new version replaces the previous 'non-LV' version (Cat # 4478378).
Key Features:
New Simple Protocol
• 96-well plate-based workflow allows easy sample management
• Only 1 required cleanup step reduces hands-on-time
• No library amplification with qPCR quantification
New Ion AmpliSeq™ HiFi Master Mix
• Enables cleaner amplification for increased coverage uniformity
• Up to 3,072-plex PCR allows larger panel designs
New Digestion Reagent
• Cleaner digest allows for more efficient downstream sequencing
Scalable Multiplex PCR Reactions
The Ion AmpliSeq™ 2.0 technology enables scalable multiplex PCR reactions from 12- to 3,072-plex in a single well using just 10 ng of starting DNA. Ion AmpliSeq™ panels and primer pools allow highly multiplexed PCR amplification of thousands of genomic target regions, with superior coverage uniformity and specificity, without the need of a specialized microfluidics PCR platform. In addition, the primers contain proprietary modifications that enable removal of primer sequences during library preparation, for efficient target assessment during sequencing. Multiple primer pools can be used to create overlapping amplicons that enable complete coverage of large targets. Ion AmpliSeq™ Custom Primer Pools are designed via the Ion AmpliSeq™ Designer web-hosted software, available at www.ampliseq.com.
Barcoded Library Preparation
The Ion AmpliSeq™ Library Kit 2.0 includes reagents for generating amplicons with Ion AmpliSeq™ primers and preparing libraries from the resulting amplicons. The kit enables the preparation of barcoded libraries using Ion Xpress™ Barcode Adapters 1–96 kits or IonCode™ Barcode Adapters 1-384 kits. Barcoded libraries can be combined and loaded onto a single Ion chip to minimize the sequencing run time and cost and allow for accurate sample-to-sample comparisons.
Easy and Flexible Protocol
The Ion AmpliSeq™ Library Kit 2.0 uses a plate-based format for easier sample handling and tracking, and for compatibility with automation and high-throughput laboratories. The library preparation is completed in as little as 3.5 hours using 10 ng of DNA per PCR, and DNA from a variety of sources—including formalin-fixed, paraffin-embedded (FFPE) tissue can be used as the starting material.
The resulting DNA libraries are ready for downstream template preparation for clonal amplification on Ion Sphere™ Particles using the automated Ion OneTouch™ System. The intuitive Torrent Suite Software and optional Ion Reporter enable you to go from extracted DNA to variant calls in as little as 10 hours.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
고처리량 호환성High-throughput Compatible, Multiplexing
라이브러리Targeted Sequencing Library
멀티플렉스 기능12- to 3072-plex per well
반응 수96 Reactions
제품라인Ion AmpliSeq
제품 유형Library Kit 2.0
수량96 reactions
샘플 종류DNA (Genomic), DNA from FFPE Tissues, Tissue (FFPE)
시퀀싱 유형Genome & DNA Sequencing
종Human
시작 물질10 ng gDNA
기술Library Generation
용도(애플리케이션)Next-Generation Sequencing (NGS)
Unit SizeEach
구성 및 보관
• 7 tubes, store at -10°C to -30°C
• 1 tube Low TE Buffer, store at room temperature
자주 묻는 질문(FAQ)
When should I consider designing my own amplicons versus using the Ion Ampliseq Designer and Ion AmpliSeq technology?
The Ion AmpliSeq Designer (www.ampliseq.com) supports custom designs for targets in the human and mouse genomes. There are two design option sizes 125-175 bp, recommended for degraded DNA samples, and 125-275 bp, recommended for standard DNA input. Ion Ampliseq technology is a simple, fast, and affordable targeted sequencing strategy based on ultrahigh-multiplex PCR. The library preparation is completed in as little as 3.5 hours using 10 ng of DNA per PCR. To learn more about Ion Ampliseq technology, please review the Ion Ampliseq FAQs.
For other genomes (non-human or non-mouse) or applications requiring longer reads (up to 400-base reads), we recommend designing your own amplicons.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the difference between a DNA fragment library and a DNA amplicon library?
A DNA fragment library is constructed from whole genomic DNA and is commonly used for whole genome resequencing or de novo sequencing. Briefly, the whole genomic DNA is fragmented or sheared, ligated with Ion-specific adapter sequences, and then size-selected for the library fragments of the desired length.
Amplicon libraries are constructed from PCR-amplified DNA fragments and are used for targeted sequencing (e.g., investigating variants at known genomic locations). There are two types of amplicon libraries, short and long.
A short amplicon library contains DNA fragments (targets) with lengths that are compatible with the Ion template preparation kits without any further shearing or fragmentation during library preparation. Additionally, no size-selection step is required, as the amplicons are already within the desired size range.
A long amplicon library contains DNA fragments (targets) with lengths that are longer than those compatible with the Ion template kits and requires further shearing or fragmentation during library preparation. The library preparation protocol for long amplicons is similar to fragment libraries.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion Library Equalizer Kit compatible with Ion Ampliseq libraries?
The Ion Library Equalizer Kit (Cat. No. 4482298) is recommended for use with Ion AmpliSeq libraries and provides an alternative to library quantification methods by using bead-based technology to normalize the final library concentration to ~100 pM.
The Ion Library Equalizer Kit is fast and cost-effective compared to traditional quantification methods; however, it may be not be the right choice for all users. Briefly, the library is amplified with the Ion Equalizer Primers, captured onto Equalizer Beads, and the normalized library is eluted from the beads using a specially formulated Equalizer Elution Buffer. The final library is normalized to ~100 pM, but there is no quality control information (e.g., measured concentration or size distribution) that can be obtained, which is possible if using the recommended library quantification kits: Ion Library Quantitation Kit (qPCR), Qubit dsDNA HS Assay Kit (Qubit 2.0 Fluorometer), or Agilent High Sensitivity DNA Kit (Agilent Bioanalyzer instrument).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
My custom design has low predicted coverage, what can I do to improve it?
If you've reviewed your design and are not satisfied with the results, please click on the “Not happy with this design? Let us help” link to have an Ion Ampliseq team member contact you about additional options for your design.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How should I dilute my library for template preparation? How much volume of the diluted library is required?
For template preparation, Ion AmpliSeq libraries are diluted to 100 pM, and the volume required for template preparation will vary depending on the template preparation kit used. Please see the Ion AmpliSeq Library Preparation User Guide for details regarding library dilutions and input into template preparation.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.