Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, extra pure
Thermo Scientific Chemicals

Ammonium iodide, 99%, extra pure

Ammonium iodide, 99%, H4IN, CAS Number-12027-06-4, unii-oz8f027ldh, ammonium iodide, hsdb 476, ammoniumiodid, ammonium iodide nh4 i, nh4i, oz8f027ldh, ammonium jodatum, azanium iodide, ammonium-iodid, 500g, 98.5% min. (Argentometry), CHEBI:63040, 2.5100g/mL, 234-717-7, 144.94, Extra Pure | CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 448075000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS12027-06-4
IUPAC Nameammonium iodide
Molecular FormulaH4IN
InChI KeyUKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES[NH4+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Titration Argentometric98.5 to 101.5 %
Residue after ignition=<0.05 %
Heavy metals (as Pb)=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only