5500 W Barcode Conversion Primers 1-16 Kit

Catalog number: 4481067

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 W Barcode Conversion Primers 1-16 Kit contains barcoded primers for libraries constructed with truncated adaptors, such as Agilent Technologies' SureSelect™ libraries, for multiplex sequencing on 5500 W Series genetic analyzers. This kit does not contain 5500 W Conversion Enzyme and should be used in conjunction with the 5500 W Conversion Primers Kit. This kit contains primers for 16 barcodes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: SOLiD™
Product Size: 48 reactions

Contents & storage

Store at -15 to -25 °C.

Documents

Manuals & protocols