MicroAmp™ Optical 384-Well GPLE Reaction Plate with Barcode
Applied Biosystems™
MicroAmp™ Optical 384-Well GPLE Reaction Plate with Barcode
Applied Biosystems MicroAmp Optical 384-Well GPLE Reaction Plates with Barcode are validated and tested for optimal fit and performance onMás información
Have Questions?
Número de catálogo 4481195
Precio (MXN)
-
Applied Biosystems MicroAmp Optical 384-Well GPLE Reaction Plates with Barcode are validated and tested for optimal fit and performance on Applied Biosystems thermal cyclers and real-time PCR instruments.
Features of the MicroAmp Optical 384-Well GPLE Reaction Plates include:
• Each lot is certified DNA-, RNase-, and PCR inhibitor-free
• Labeled “For Laboratory Use”
• Optimized for performance with Applied Biosystems instruments
• Excellent heat transfer from thermal block to sample with thin-wall polypropylene wells
• Raised deck for efficient plate sealing to help prevent evaporation and minimize contamination
MicroAmp Optical 384-well reaction plates are constructed from a single rigid piece of polypropylene and include a raised deck for efficient plate sealing. The full-skirted plate provides stability for sample loading and handling. The thin-wall polypropylene wells provide excellent heat transfer from the thermal block to the sample for fast, efficient PCR amplification.
With General Purpose Lab Equipment (GPLE) plastics, each lot of reaction plates is certified in an ISO 13485-registered facility to be free of DNA, RNase, and PCR inhibitors. GPLE plastics are ideal for use in diagnostic procedures.
Especificaciones
Línea de productosMicroAmp™
ColorOptical
Tipo de productoMicroplate
Para utilizar con (equipo)3130xl Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3730xl DNA Analyzer, 7900 HT System, GeneAmp 9700, QuantStudio™, Veriti Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, QuantStudio™ Dx, 3130 Genetic Analyzer, 3500 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Dx Genetic Analyzer
Estilo de faldaFull
MaterialPolypropylene
Volumen del pocillo (métrico)40 μL
N.° de pocillos96
Cantidad50 plates
Unit Size50 Plates