Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals

Have Questions?
產品號碼QuantityPackaging
448182500250 gPlastic bottle
產品號碼 448182500
價格 (TWD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS7782-75-4
IUPAC Namemagnesium(2+) trihydrate hydrogen phosphate
Molecular FormulaH7MgO7P
InChI KeyOKIWLDVQGKRUNR-UHFFFAOYSA-L
SMILESO.O.O.[Mg++].OP([O-])([O-])=O
檢視更多
規格Specification Sheet規格表
Titration Complexometric>=97.5 %
Solubility(0.1 M in 5 M HCl) Clear colorless
Heavy metals (as Pb)=<20 ppm
UV(0.1 M in 5 M HCl)
UVat 260 nm A: =<0.035
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only