Ion Torrent™

Covaris™ M220 microTUBE, Screw-Cap

Catalog number:  4482264

 Related applications:

Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Covaris® M220 Screw-Cap microTUBE has been designed and engineered specifically for the treatment of 50 µl of sample in the M220 Focused-ultrasonicator™. The microTUBE is used to shear DNA and RNA into fragments with a distribution center selected between 100 to 1,500 bp. Each package contains 25 tubes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Product Line: Covaris®
Product Size: 25⁄box
Tube Type: DNA/RNA Shearing Tube
Type: Tubes & Vials

Contents & storage

Store at room temperature

Documents

Manuals & protocols