Covaris® M220 miniTUBE Prep Station & Stand

Catalog number: 4482267

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Covaris® M220 miniTUBE Prep Station enables the easy loading of sample into clear, blue, or red miniTUBEs. A tilted stand is included that has been designed to help recover the entire sample from the miniTUBE. Each package contains 1 preparation station.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Product Line: Covaris®
Product Size: 1 each
Type: Stand

Contents & storage

Store at room temperature

Documents

Manuals & protocols