Ion Torrent™

Covaris™ M220 miniTUBE, Blue

Catalog number:  4482268

 Related applications:

Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Covaris® M220 Blue miniTUBE is specifically engineered to shear genomic DNA into fragment size distributions centered on 3 kb. Fragment distributions are tight and precisely centered on the targeted size. Each blue miniTUBE can process 200 µl of sample. Each package contains 25 tubes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Product Line: Covaris®
Product Size: 25⁄box
Tube Type: DNA/RNA Shearing Tube
Type: Tubes & Vials

Contents & storage

Store at room temperature

Documents

Manuals & protocols