Covaris® M220 microTUBE Holder

Catalog number: 4482279

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Covaris® M220 microTUBE Holder is required to process the sample tube in the M220 Focused-ultrasonicator™. This holder is compatible with snap-cap and screw-cap microTUBEs. Each package contains 1 holder station.
For Research Use Only. Not for use in diagnostic procedures.


Type: Holder
Product Size: 1 kit
For Use With (Equipment): Ion PGM™ System

Contents & storage

Store at room temperature


Manuals & protocols