Covaris® M220 AFA-grade Water

Catalog number: 4482280

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Highly purified water (at least ASTM Type III or ISO Grade 3) is required when using the M220 Focused-ultrasonicator™. One AFA-grade water bottle (800 ml) is sufficient for more than 50 uses.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity or Quality Grade: Qualified
Sample Treatment: None
Format: Bottle(s)
Product Line: Covaris®
Product Size: 1 each
Quantity: 1 bottle(s)

Contents & storage

Store at room temperature

Documents

Manuals & protocols