TaqMan™ QSY and QSY2 Probes
Applied Biosystems™

TaqMan™ QSY and QSY2 Probes

Applied Biosystems TaqMan QSY and QSY2 probes are dual-labeled probes used for real-time PCR. TaqMan QSY and QSY2 Probes are designed for optimal sensitivity and performance in multiplexed qPCR experiments for detection of up to 6 targets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
4482777QSY Probe6000 pmol
4482778QSY Probe20,000 pmol
4482779QSY Probe50,000 pmol
A56899QSY2 Probe6000 pmole
A56900QSY2 Probe20,000 pmole
A56901QSY2 Probe50,000 pmole
Catalog number 4482777
Price (USD)
346.65
Online exclusive
365.00
Save 18.35
Each
Product Type:
QSY Probe
Quantity:
6000 pmol

Applied Biosystems TaqMan QSY and QSY2 probes are dual-labeled probes used for real-time PCR with Cyanine 5 (Cy5) and Cyanine 5.5 (Cy5.5) reporter dyes. TaqMan QSY and QSY2 Probes are designed for optimal sensitivity and performance in multiplexed qPCR experiments for detection of up to 6 targets. Current users of dark quencher probes, such as Black Hole Quencher (BHQ™) probes, can design custom QSY and QSY2 probes without changing probe sequences.

All TaqMan QSY and QSY2 probes are HPLC-purified and verified by mass spectrometry to help ensure lot-to-lot reproducibility between batches. Thermo Fisher Scientific has been a leader in qPCR probe manufacturing for over 20 years, with expertise in probe synthesis in a facility that is certified to ISO 13485. TaqMan QSY and QSY2 probes can be used for viral pathogen and microbial detection in research samples for healthcare and biopharmaceutical development needs.

Features of TaqMan QSY and QSY2 probes include:

  • Cyanine 5 (Cy5) and Cyanine 5.5 (Cy5.5) reporter dyes—new TaqMan QSY2 probes include Cy5 and Cy5.5 reporter dyes, now enabling you to multiplex up to six targets
  • Increased sensitivity compared to probes from another leading supplier—detect low-abundance targets, impurities, and fluctuations in bioproduction more accurately
  • Multiplexing performance without compromise—use QSY for multiplexing up to four targets with ABY, JUN, FAM, and VIC and use QSY2 for 5th and 6th target multiplexing with Cy5 and Cy5.5
  • Better performance retention when scaling up—consistent performance from singleplex to 6-plex creates a smoother assay development experience
  • TaqMan QSY and QSY2 probes have greater dynamic range compared to probes from another leading supplier—enables more accurate detection of a greater range of analyte concentrations
  • Get started quickly—BHQ probe sequences can be converted to QSY and QSY2 versions with no redesign needed
  • Compatible with our master mixes and validated on our instruments—get increased confidence in the performance of the probes in your lab to help get your products to market faster
  • Part of the Thermo Fisher Scientific qPCR ecosystem—our teams can support you in all components of the real‐time PCR (qPCR) workflow

Guaranteed yield

The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount. Ensure that you are considering the guaranteed yield (amount of probe you are actually receiving) when calculating price per reaction.

Approximate ship times

  • 4 to 10 days in North America
  • 5 to 10 days in Europe

Calibration

For calibration of ABY, JUN, and Cy5, the complete list of calibration plates for 96-well, 0.1 mL; 96-well, 0.2 mL; and 384-well plate formats are in the Multiplexing Optimization Guide in the document section. For Calibration of Cy5.5, please email your request to specialty_oligos@thermofisher.com.

We also offer Applied Biosystems custom primers and other types of TaqMan probes.

Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs.

Specifications
3'Primer ModificationQSY
5'Primer ModificationABY, FAM, JUN, VIC
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
FormatTube
Internal Probe ModificationNone
Product LineTaqMan
Product TypeQSY Probe
Purification MethodHPLC
Quantity6000 pmol
Shipping ConditionRoom Temperature
Detection MethodPrimer-probe
FormLiquid or Dried Down
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a dual-labeled probe in solution or dried down.

Store at -15°C to -25°C

Frequently asked questions (FAQs)

I need to see all the primer and probe sequences from an assay design right away. How can I do this?

You can directly see all the primer and probe sequences from a test design right away using the Primer Express Software. Open the software and choose File => New. Choose either TaqMan MGB Quantification or TaqMan Quantification for Gene Expression Assay design. Copy/paste your gene sequence of interest into the new window and click on the green triangle to get primer/probe designs.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How can I determine whether the Tm of my custom probe is appropriate?

In general, the target Tm of a TaqMan probe is 70 degrees C. Using the Primer Express Software, you can check the Tm of a MGB or TAMRA probe probe.

Within the software, go to Primer Probe Test Tool and then copy/paste in your sequence of interest to see the Tm. If you need to check for a MGB based probe, make sure to choose Document Type: TaqMan MGB Quantification. If you need to check for a QSY or TAMRA based probe, make sure to choose Document Type: TaqMan Quantification. Real-Time PCR and Digital PCR Applications

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

I ordered a VIC-QSY–labeled TaqMan probe, however, I received probes with a QSY7 label. Is this correct?

QSY7 is the QSY quencher that we use in TaqMan probes. You can go ahead and use this probe in your qPCR application.

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.

Do I need to redesign my probe when converting from BHQ and other competitor probes to TaqMan QSY/QSY2 Probes?

No, redesign is not necessary when converting from most competitor probes to TaqMan QSY/QSY2 Probes.

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.

Is there a need to adjust filter settings on the instrument to utilize Cyanine 5 and Cyanine 5 dyes with TaqMan QSY/QSY2 Probes?

Yes, please make sure to enable the x6m6 filter (6th channel) in the instrument filter settings prior to the qPCR run. This filter is not preselected by default.

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.