Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ EnduraPlate™ Optical 384-Well Blue Reaction Plates with Barcode
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 384-Well Blue Reaction Plates with Barcode

Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs.Read more
Have Questions?
Catalog NumberQuantity
448332020 plates
Catalog number 4483320
Price (USD)
204.65
Online exclusive
218.00
Save 13.35
Each
Quantity:
20 plates
Price (USD)
204.65
Online exclusive
218.00
Save 13.35
Each
Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. All of our plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. With the new EnduraPlate™ reaction plates, there is now a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments.

Key Features and Benefits:

• Optimized for performance with Applied Biosystems™ instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs

Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability. ANSI/SBS compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler -> real-time thermal cycler -> genetic analyzer).

EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity, while five color choices provide a convenient way to organize your laboratory assays. A GPLE (For Laboratory Use) version is also available.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging20 Plates
Product TypeMicroplate
Quantity20 plates
Reaction SpeedStandard
Skirt StyleSkirted
BarcodeIncludes Barcode
ColorOptical, Blue
MaterialPolycarbonate, Polypropylene
No. of Wells384
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Store at room temperature.