Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles
Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantity
448361000100 mL
Catalog number 448361000
Price (USD)
70.10
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
70.10
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.80 %
Filt.over 0.2 micron PFTEFiltered
Water=<0.005 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only