Ridom™ SeqSphere+ Software for Ion Torrent™ Systems (Commercial)
Product Image
Ion Torrent™
Ridom™ SeqSphere+ Software for Ion Torrent™ Systems (Commercial)
SeqSphere+ Software from Ridom allows for automatic processing and analyzing of microbial sequence data obtained from Ion Torrent™ systems. SeqSphere+Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLicense
4483696Commercial, 3 Year, 2 Seat
4483692Commercial, 1 Year, 2 Seat
4483700Commercial, 5 Year, 2 Seat
4483694Commercial, 1 Year, 5 Seat
4483698Commercial, 3 Year, 5 Seat
4483702Commercial, 5 Year, 5 Seat
Catalog number 4483696
Price (MXN)
-
License:
Commercial, 3 Year, 2 Seat
SeqSphere+ Software from Ridom allows for automatic processing and analyzing of microbial sequence data obtained from Ion Torrent™ systems. SeqSphere+ Software enables whole genome microbial typing (MLST+) or traditional MLST sequencing projects. This software is designed both for individual labs and for distributed work-groups (client/server model) to enable easy data sharing. License options are available for 1, 3, or 5 years; 2 or 5 seats; and commercial or academic/government labs.
Ridom SeqSphere+ Software Features:
User-friendly: no scripting skills or bioinformatician needed
Highly automated workflow: download a pre-designed task template or create a custom template, then apply it to the analysis of hundreds of strains with almost no user intervention.
Single, expanding nomenclature service: worldwide, unique nomenclature service that enables a 'Molecular Typing Esperanto'
DNA re-sequencing editor: edit and analyze reference-mapped assemblies of Ion Torrent™ data (e.g., MLST, MLST+) with auto-correction of homopolymer-related insertion/deletion errors.
Sanger data: assemble, edit, and analyze Sanger CE sequencing data (e.g., MLST).
Analytical tools: select data entries from a comparison table for epidemiologic, evolutionary, or functional analysis. Cluster and visualize data entries by using minimum spanning or UPGMA/neighbor-joining trees.
Database:
   --Store, search, retrieve, export, and create reports from your epidemiologic and DNA sequence data stored in an integrated database. Search new sequence entries against stored data.
   --Data fields are compliant with the metadata requirements of the NCBI BioSample. Enables data entry plausibility checks.
Bacterial typing: typing of bacteria is automatically performed with user-defined quality parameters (e.g., coverage, stop codons) using public or self-defined query libraries and task templates.
Ridom Community:
   --Rapidly and easily share task templates with other institutions or download them online from the Task Template 'Store'.
   --Option to contribute to a single, worldwide, expanding, publicly-available database of nomenclature and epidemiologic data. Larger (supra) national institutions may obtain their own server for epidemiologic data upon request.
Security:
   --Encryption (SSL) of data in transmission
   --Various configurable user roles, user groups, and access controls
   --Audit trail functionality (who, when and what)
Computer Specifications (Minimum)
Client computer OS: Windows™ 7 64-bit, CPU: Core i5, RAM: 8 GB, HD: 500 GB, Internet connection required to use public Ridom MLST+ Nomenclature Server & Task Template 'Store' (supports working with a private and local nomenclature and to generate custom tasks).
Server computer OS: Windows™ 7/Linux 64-bit (Ubuntu LTS recommended), CPU: Core i5, RAM: 8 GB (48 GB recommended if MIRA de novo assembly done on this server), HD: 2 TB (RAID level 1 or 5 recommended), it is recommended not to use the NGS machine computer as server.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Sequencing Data Analysis
LicenseCommercial, 3 Year, 2 Seat
Operating SystemIon Personal Genome Machine (PGM™) System
Product LineRidom™ SeqSphere+
Product TypeSeqSphere+ Software
Quantity3 years-2 users
Unit SizeEach