This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now

TaqMan® Cronobacter spp Assay Beads

Catalog number: 4485035

 Related applications: Food & Beverage

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These TaqMan® assay beads provide a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viral target. The assay uses PCR to amplify a DNA target unique to the microorganism and TaqMan® probe to detect and quantify the target, all in an an easy-to-use, lyophilized bead format. TaqMan® assay beads deliver the fastest RT-PCR run times on the market (<40 minutes in Fast mode). Simply add 30 µL of purified DNA to the beads and run on Fast or standard instrumentation. These TaqMan® assay beads provide microbial detection capabilities and high-confidence results to help accelerate your food and environmental testing needs.

Features of TaqMan® assay beads:

• Convenience—simply add your nucleic acid sample to the bead-containing assay tube and run; no pre-mixing or master mix preparation required
• Consistency—fewer operator steps means reduced error and increased reproducibility across time, users, and laboratories
• Improved data accuracy—an internal positive control is included to monitor your PCR
• Fast chemistry—40-min run time with Fast cycling, compatible with standard or Fast modes

TaqMan® assay beads are suitable for single or multiplex analysis. Because they are run using universal conditions, multiple bead-based assays can be analyzed in the same run. The robust assay design provides highly accurate, reproducible, and reliable results, with the enhanced stability of a lyophilized format. A passive internal reference based on proprietary ROX™ dye is included for precise data analysis.

This product is for environmental testing, quality control/quality assurance testing, food and agricultural testing, and research purposes only.
For Testing of Food and Environmental Samples Only.

Specifications

Comments: Includes Master Mix
Form: Lyophilized
Target Organism Class: Cronobacter spp.
Validated Application: Polymerase Chain Reaction
Detection Method: Primer-Probe Detection
Reaction Speed: Fast, Standard
Product Size: 96 reactions
Species: All Species

Contents & storage

• 12 8-tube strips containing assay beads (store at 2–8°C in the dark)
• Optical 8-tube strips and 8-cap strips (store at room temperature)

These reagents are sufficient for 96 reactions.

Documents

Manuals & protocols