This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

96-Well Pre-Clarification Tray

Catalog number:  4485076

 Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 96-Well Pre-Clarification Tray is designed for use with MagMAX™ Express-96 Deep Well Plates in food safety and environmental monitoring applications. The pre-clarification tray offers a simple, cost-effective way to filter up to 96 samples at the same time from more difficult starting matrices such as pet food, fine grains, etc. The tray is designed as a filtration step before bacterial lysis and subsequent nucleic acids extraction steps. During the filtration process, the media inside the spin column removes PCR inhibitors and carryover food particles from the sample, allowing the bacteria to pass through into the 96 deep well, sample collection plate. The lysis is then ready to proceed to nucleic acid purification with, for example, the PrepSEQ® Nucleic Acid Extraction Kit for Food and Environmental Testing.
For Testing of Food and Environmental Samples Only.

Specifications

Color: Clear
For Use With (Equipment): MagMAX™ Express-96 Deep Well Magnetic Particle Processor
Plate Compatibility: 96-Well Plates
Product Size: 1 plate
Shipping Condition: Room Temperature
Type: Filter

Contents & storage

Store at room temperature. Expiration date is five years from date of manufacture.

Documents

Manuals & protocols

Recommended products