Ion Torrent™
Partek™ Flow™ Software, 12 month license (Government)
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Partek® Flow® Software is specifically designed for front-end alignment, enhanced quality control, RNA quantification, and variant detection analysis of next-generation sequencing data as part of your research. Use it alone, or combined with Partek® Genomics Suite™ Software, for complete start-to-finish next generation sequencing data analysis (see figure below for a comparison of features). This product, which can be ordered through our website, is supplied directly by Partek Incorporated.
• Get started right away with a user-friendly interface that guides selection and alignment and organizes work into projects for intuitive navigation
• Easily add users to collaborate and share protocols and data within your institution to save time and leverage collective knowledge
• Schedule tasks to run when resources become available and receive e-mail notifications when tasks are complete so you can focus your efforts where they are needed most
• Desktop, server, cluster, and cloud-friendly installation allow for multi-user site license option, the ability to effortlessly process enormous data sets, and the ability to access your data from any browser, anywhere
• Process raw data from all leading next-generation sequencing technologies
• Automatically record your project history for easy auditing and tracking
• Save storage space and reduce your file output size and transfer time by automatically converting to BAM files and compressed project files that seamlessly transition data to Partek® Genomics Suite™ Software
• Supports all core assays allowing you to analyze data using a variety of approaches
This Partek® Flow® Software product is manufactured and offered by Partek Incorporated. Purchases of this product will be processed by us on behalf of Partek, as will all subsequent fulfillment, shipment, and support needs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): Ion PGM™ System, Ion Proton™ System
Software Type: Sequencing Data Analysis
Data Input: BAM, FASTQ, SAM, SFF
License Type: Government
Operating System: CentOS, Fedora, Red Hat, Ubuntu, Windows 7, Windows 8, Windows XP SP3
Product Line: Partek®
Product Size: 1 each
Software Category: Software
Documents
Manuals & protocols