Partek® Flow® + Genomics Suite™ Software Bundle, 12 month license (Commercial)
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Partek® software suite is a complete start-to-finish genomic data analysis solution, providing powerful statistics and visualizations. Start with raw NGS reads from the Ion PGM™ or Ion Proton™ sequencers and quickly identify biologically meaningful results and pathways. The user-friendly point and click interface provides streamlined analysis to guide the novice user without sacrificing advanced functionality.
This bundle combines both the front-end Partek® Flow® pipeline manager and the downstream Partek® Genomics Suite™ analysis tools to provide a complete end-to-end solution for RNA-Seq applications. (See figure below for a comparison of features.)
This Partek® Flow® + Genomics Suite™ bundle is manufactured and offered by Partek Incorporated. Purchases of this product will be processed by us on behalf of Partek, as will all subsequent fulfillment, shipment, and support needs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): Ion PGM™ System, Ion Proton™ System
Software Type: Sequencing Data Analysis
Data Input: BAM, FASTQ, SAM, SFF
License Type: Commercial
Operating System: CentOS, Fedora, Red Hat, Ubuntu, Windows 7, Windows 8, Windows XP SP3
Product Line: Partek®
Product Size: 1 each
Software Category: Software
Documents
Manuals & protocols