Sodium borohydride, (VenPureTM solution), 12% aq. solution
Sodium borohydride, (VenPureTM solution), 12% aq. solution
Sodium borohydride, (VenPureTM solution), 12% aq. solution
Thermo Scientific Chemicals

Sodium borohydride, (VenPureTM solution), 12% aq. solution

CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 448511000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Iron (Fe)=<10 ppm
Methanol=<2000 ppm
Appearance (Form)Clear liquid
Titration with HCl38.0 to 42.0 % w/w (NaOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only