Search
View additional product information for Ion PI™ Template OT2 200 Kit v2 - FAQs (4485146)
14 product FAQs found
The "time remaining" is not entirely accurate. The centrifuge may keep rotating for about 15 minutes after the time has become 0:00. If it rotates for more than 15 minutes, please contact Technical Support.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The pressure inside the instrument may be above normal. If the Ion OneTouch 2 firmware is older than v540, please update it to the latest version.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
It may be rubber from the packing. The amount varies among instruments and it is not anything to be concerned about, as it will be removed by the Ion OneTouch ES.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
No, you cannot. Please exchange the oil when switching between different kits. Please contact Technical Support for detailed information when switching kits.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Open the chip clamp and check for any leaks. If you see solution leaking, please remove the chip and clear the solution with a Kimwipe. DO NOT wipe or scrub as the socket could be damaged. Clean the socket by gently touching it with a damp Kimwipe. Make sure the socket is dry and replace the chip. Press the 'Calibration' button to recalibrate the chip. If it fails again, change the chip to a different one. Also, if you are using an Ion 316 Chip v2 on a v3.4 or older instrument, the instrument will not recognize the chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The current Ion PI Template OT2 200 Kit v2 (Cat. No. 4485146) and Ion PI Sequencing 200 Kit v2 (Cat. No. 4485149) support up to 200-base reads (~200 bp median insert size, or ~270 bp median library size).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Firmware updates to the software controlling the Ion OneTouch 2 Instrument are periodically released. To update the firmware to the appropriate version, use either a USB flash drive or an ethernet connection, as described on page 11 of the Ion OneTouch 2 System - Installation, Setup, and Maintenance User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0014388_IonOneTouch2Sys_UG.pdf ).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
We recommend removing the samples from the Ion OneTouch 2 instrument ≤ 16 hours after starting the run. After centrifuging and removing the samples from the instrument, follow the ISP collection protocol to recommended stopping points and follow the instructions for preparing the unenriched ISPs for storage. The ISP suspension can be stored at 2 degrees C to 8 degrees C for up to 3 days.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
In a typical Ion OneTouch 2 run, you should expect ~18 mm drop in the level of oil in the tube (~22 mL consumed), and ~13 mm drop in the recovery solution (~15 mL consumed). During cleaning, you should expect to collect ~10 mL of oil in the 50 mL conical waste collection tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Due to differences in reagent formulations between kit types, we recommend following the Demonstrated Protocol: How to Set Up the Ion OneTouch 2 Instrument When Switching Between the Ion PGM System and Ion Proton System (http://ioncommunity.thermofisher.com/docs/DOC-7265) if using more than one Ion OneTouch 2 Template kit type on your instrument. Briefly, the instrument lines must be primed with the new kit reagents by running a cleaning and initialization procedure prior to setting up the next run. Please see the above user guide for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The input amount for template preparation on the Ion OneTouch 2 is library type specific. Occasionally, library input titration may be required to optimize templated ISP clonality and maximize sequencing throughput. Please see the specific guidelines for your library type in the appropriate library preparation user guide that can be found here: http://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/next-generation-sequencing-support-center/ngs-library-preparation-support.html
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
For the Ion PGM System, we offer:
- Ion PGM Hi-Q View OT2 Kit (Cat. No. A29900) for template preparation of libraries up to 400 bp in length.
For the Ion Proton System, we offer:
- Ion PI Hi-Q OT2 200 Kit (Cat. No. A26434) for template preparation of libraries up to 200 bp in length.
For the Ion S5 System and Ion S5 XL System, we offer two kits:
- Ion 520/530 Kit-OT2 (Cat. No. A27751) for template preparation of libraries up to 400 bp in length.
- Ion 540 Kit- OT2 (Cat. No. A27753) for template preparation of libraries up to 200 bp in length.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
In a typical Ion OneTouch 2 run, you should expect ~18 mm drop in the level of oil in the tube (~22 mL consumed), and ~13 mm drop in the recovery solution (~15 mL consumed). During cleaning, you should expect to collect ~10 mL of oil in the 50 mL conical waste collection tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Ion Torrent OneTouch2 protocols can be found at our NGS Support Center (thermofisher.com/ngssupport), in the NGS Template Preparation Support Center section. For all other protocols, please go here (http://www.thermofisher.com/manuals-protocols-search.html).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.