2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals
2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals
2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals

CAS: 51285-26-8 | Molecular Formula: C6H7N3·ClH | Molecular Weight: 157.6 g/mol
Have Questions?
Catalog NumberQuantity
4486100505 g
Catalog number 448610050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS51285-26-8
IUPAC Name[amino(pyridin-2-yl)methylidene]azanium
Molecular FormulaC6H8N3
InChI KeyKNXKVYCVGXFLES-UHFFFAOYSA-O
SMILESNC(=[NH2+])C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
HPLC>=96.0 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration Argentometric>=96.0 to =<104.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only