Diagenode™ iDeal ChIP Kit

Catalog number: 4487119

Ion Torrent™  Related applications: Chromatin Biology | Epigenetic Sequencing | Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Diagenode™ iDeal ChIP-seq Kit provides optimized reagents and protocols for successful chromosomal immunoprecipitation followed by next-generation sequencing (ChIP-seq), significantly reducing the time and effort typically required for the optimization of reaction conditions. The iDeal ChIP-seq kit has been designed for high-quality ChIP followed by sequencing on virtually any platform. The kit has been validated for use with the major commercial sequencers, including the Ion PGM™ and Ion Proton™ instruments.

The complete kit includes control antibodies, control primers, and a DNA purification module. Diagenode’s long history of developing optimal epigenetics tools and our expertise with ChIP-seq enables reproducible, efficient, and high quality results every time.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System, Ion Proton™ System
Library Type: Fragment Library
Workflow Step: Library Generation
Starting Material: DNA
Sequencing Type: Epigenetic Sequencing
Comments: This 3rd party vendor product will be drop shipped to the customer from the vendor.
Product Line: Diagenode™
Product Size: 24 reactions

Documents

Manuals & protocols

Recommended products