Ion AmpliSeq™ Library Kit Plus
Ion Torrent™

Ion AmpliSeq™ Library Kit Plus

The Ion AmpliSeq Library Kit Plus is designed for rapid preparation of amplicon libraries using Ion AmpliSeq panels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
448899024 Reactions
A3590796 Reactions
A38875384 Reactions
Catalog number 4488990
Price (MXN)
-
No. of Reactions:
24 Reactions

The Ion AmpliSeq Library Kit Plus is designed for rapid preparation of amplicon libraries using Ion AmpliSeq panels. Ion AmpliSeq technology enables scalable, multiplex polymerase chain reactions (PCR) from 12- to 24,000-plex in a single well. Next-generation sequencing (NGS) libraries can be prepared using as little as 1 ng of starting material.

Features of the Ion AmpliSeq Library Kit Plus include:

  • Scalable and efficient library preparation
  • Easy and robust protocol
  • Barcoded sample multiplexing

Scalable and efficient library preparation
Using as little as 1 ng of starting material, highly multiplexed PCR amplification of thousands of genomic target regions yields superior coverage uniformity and specificity, without the need of a specialized microfluidics PCR platform. The primers contain proprietary modifications that enable removal of primer sequences during library preparation, thereby resulting in efficient target assessment during sequencing.

Easy and robust protocol
The Ion AmpliSeq Library Kit Plus uses a plate-based format for easier sample handling and tracking, and for compatibility with automation and high-throughput laboratories. The protocol is simple to use and enables high yield and uniformity, reliably and reproducibly. The resulting libraries are ready for downstream template preparation using the automated Ion Chef System (which also provides the option of automated library preparation using the Ion AmpliSeq Kit for Chef DL8 (Cat. No. A29024)) or the Ion OneTouch 2 System followed by sequencing. The intuitive Torrent Suite Software and optional Ion Reporter Software enables automation of data analysis.

Barcoded sample multiplexing
The Ion AmpliSeq Library Kit Plus includes reagents for generating amplicons using Ion AmpliSeq primers and preparing libraries from the resulting amplicons. The kit enables the preparation of barcoded libraries using Ion Xpress Barcode Adapters 1-96 kits or IonCode Barcode Adapters 1-384 kits. Barcoded libraries can be combined and loaded onto a single Ion chip to minimize the sequencing run time and cost, and to allow for accurate sample-to-sample comparisons.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
LibrariesTargeted Sequencing Library
No. of Reactions24 Reactions
Product LineIon AmpliSeq
Product TypeLibrary Kit Plus
Quantity24 reactions
Sample TypeDNA
Shipping ConditionApproved for shipment on Wet or Dry Ice
FormatKit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between the Ion AmpliSeq Library Kit Plus (Cat. No. 4488990) and the Ion AmpliSeq Library Kit 2.0 (Cat. No. 4475345)?

Both kits are used for the same function (preparation of amplicon libraries using Ion AmpliSeq panels). The difference is that the Plus kit is a technical improvement in relation to the 2.0 kit. It has a different and optimized formulation that achieves slightly better results with difficult samples.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What do I need to do to prep the barcode adapters in the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

When handling barcode adapters, be careful to avoid cross-contamination by changing gloves frequently and by carefully removing the plate seal.
- Pulse vortex the IonCode Barcode Adapters plate several times.
- Centrifuge the IonCode Barcode Adapters plate for 1 minute at 1000 x g to collect contents.
- Carefully remove, then discard the plate seal.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many reactions are possible with the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit consists of 1 x 96-well plate, with 20 µL of each barcode per well. The plate consists of 960 reactions in total with 10 reactions for each of the 96 barcodes.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

In what format are the barcode adapters in the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit consists of 1 x 96-well plate, with 20 µL of each barcode per well. The barcodes are pre-diluted, ready-to-use. The 96-well plate format allows multichannel pipetting for more efficient work.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the shipping and storage conditions for IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit is shipped on dry ice and we recommend storing at -30 degrees C to -5 degrees C in an upright position.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.