Proteinase K for MagMAX™-96 DNA Multi-Sample Kit
Invitrogen™

Proteinase K for MagMAX™-96 DNA Multi-Sample Kit

This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made available separatelyRead more
Have Questions?
Catalog NumberQuantity
448911096 preps
Catalog number 4489110
Price (ZAR)
-
Quantity:
96 preps
This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made available separately for applications that require more proteinase K enzyme than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeGenomic DNA
For Use With (Application)Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
FormLiquid
High-throughput CompatibilityHigh-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Product LineMagMAX
Product TypeDNA Multi-Sample Kit
PurityDNase-Free
Quantity96 preps
TargetGenomic DNA
Unit SizeEach
Contents & Storage
1 tube

Store at -25°C to -15°C.