PK Buffer for MagMAX™-96 DNA Multi-Sample Kit
Invitrogen™

PK Buffer for MagMAX™-96 DNA Multi-Sample Kit

This is the PK Buffer found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made available separatelyRead more
Have Questions?
Catalog NumberQuantity
448911111 mL
Catalog number 4489111
Price (USD)
52.25
Each
Quantity:
11 mL
Price (USD)
52.25
Each
This is the PK Buffer found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made available separately for applications that require more PK Buffer than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeGenomic DNA
For Use With (Application)Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
High-throughput CompatibilityHigh-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions96 Preps
Product LineMagMAX
Product TypeDNA Multi-Sample Kit
PurityDNase-Free
Quantity11 mL
Sample TypeBlood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
TargetGenomic DNA
Test Time1 hr.
Format96-well Plate
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
1 bottle

Store at room temperature.