DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit
Invitrogen™

DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit

These are the DNA Binding Beads found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made availableRead more
Have Questions?
Catalog NumberQuantity
448911296 preps
Catalog number 4489112
Price (USD)
130.00
Each
Quantity:
96 preps
Price (USD)
130.00
Each
These are the DNA Binding Beads found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. Nos. 4413020, 4413021, 4413022), made available separately for applications that require more DNA Binding Beads than are provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeGenomic DNA
For Use With (Application)Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
High-throughput CompatibilityHigh-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Product LineMagMAX
Product TypeDNA Binding Bead
PurityDNase-Free
Quantity96 preps
Sample TypeBlood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
TargetGenomic DNA
Test Time1 hr.
Format96-well Plate
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
1 tube, store at 2–8°C, DO NOT FREEZE.

Frequently asked questions (FAQs)

Can I purchase the RNA Binding beads from the MagMAX MirVana Total RNA Isolation Kit (Cat. No. A27828) separately?

Yes, you can. The DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4489112) are the same as that used in A27828.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.