Furosemide, 97+%
Furosemide, 97+%
Furosemide, 97+%
Thermo Scientific Chemicals

Furosemide, 97+%

CAS: 54-31-9 | C12H11ClN2O5S | 330.74 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 448970050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54-31-9
IUPAC Name4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
Molecular FormulaC12H11ClN2O5S
InChI KeyZZUFCTLCJUWOSV-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
HPLC>=97.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only