Atenolol, 98%
Atenolol, 98%
Atenolol, 98%
Thermo Scientific Chemicals

Atenolol, 98%

CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 449550050
Price (USD)
169.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
169.00
Each
Chemical Identifiers
CAS29122-68-7
IUPAC Name2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
Molecular FormulaC14H22N2O3
InChI KeyMETKIMKYRPQLGS-UHFFFAOYNA-N
SMILESCC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Melting point151°C to 155°C
Appearance (Form)Powder
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only