eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate
Invitrogen™

eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45-4317-8025 μg
45-4317-82100 μg
Catalog number 45-4317-80
Price (KRW)
240,000
Online offer
Ends: 31-Dec-2026
282,000
Save 42,000
Each
Quantity:
25 μg
Price (KRW)
240,000
Online offer
Ends: 31-Dec-2026
282,000
Save 42,000
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

695 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only
Specifications
Concentration0.2 mg/mL
Label or DyePerCP-Cy5.5
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity25 μg
Target MoleculeBiotin
ConjugatePerCP-Cy5.5
FormLiquid
Product LineeBioscience
Unit SizeEach