4-Bromo-2-methoxypyridine, 97%
4-Bromo-2-methoxypyridine, 97%
4-Bromo-2-methoxypyridine, 97%
Thermo Scientific Chemicals

4-Bromo-2-methoxypyridine, 97%

CAS: 100367-39-3 | C6H6BrNO | 188.02 g/mol
製品番号(カタログ番号) 450000050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100367-39-3
IUPAC Name4-bromo-2-methoxypyridine
Molecular FormulaC6H6BrNO
InChI KeyYFTGMMXMLPTTAY-UHFFFAOYSA-N
SMILESCOC1=NC=CC(Br)=C1
さらに表示
Infrared spectrumConforms
Refractive index1.5540 to 1.5560 (20°C, 589 nm)
GC>=96.0 %
Appearance (Color)Colorless to yellow
Appearance (Form)Low melting solid or liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only