Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000-180,000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000-180,000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000-180,000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000-180,000

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 450150250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Total volatile=<5 % (including water)
Appearance (Form)Granular solid
pH4.5 to 6.5 (4 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only