Vanadium(V) oxide, 99+%
Vanadium(V) oxide, 99+%
Vanadium(V) oxide, 99+%
Thermo Scientific Chemicals

Vanadium(V) oxide, 99+%

CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 450162500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow
Assay>=99.0 %
Phosphorus (P)=<0.04 %
Particle size=<1.2 mm
Iron (Fe)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only