2-Iminothiolane Hydrochloride, 98%
2-Iminothiolane Hydrochloride, 98%
2-Iminothiolane Hydrochloride, 98%
Thermo Scientific Chemicals

2-Iminothiolane Hydrochloride, 98%

CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 450180010
Price (USD)
244.65
Special offer
Online exclusive
Ends: 28-Dec-2026
288.00
Save 43.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
244.65
Special offer
Online exclusive
Ends: 28-Dec-2026
288.00
Save 43.35
Each
Chemical Identifiers
CAS4781-83-3
IUPAC Namehydrogen thiolan-2-imine chloride
Molecular FormulaC4H8ClNS
InChI KeyATGUDZODTABURZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].N=C1CCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration Argentometric>=97.5 %
Melting point193°C to 198°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only