MiniCollect™ Tube, Light-Blue Cap, 9NC Coagulation Sodium Citrate 3.2%, Sterile
Greiner Bio-One™
MiniCollect™ Tube, Light-Blue Cap, 9NC Coagulation Sodium Citrate 3.2%, Sterile
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
4505391 mLPack of 40
450539PK
also known as 450539
1 mLEach
Catalog number 450539
Price (USD)
-
Capacity (Metric):
1 mL
Unit Size:
Pack of 40
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
  • This light blue top sodium citrate tube, or coagulation tube, is used for examination of coagulation parameters.
  • Coagulation tubes contain a buffered sodium citrate solution.
Specifications
Closure ColorLight Blue
Closure IncludedYes
Quantity500/Pk.
Additive9NC Sodium Citrate 3.2%
Capacity (Metric)1 mL
SterilitySterile
Unit SizePack of 40