4-(N,N-Dimethyl)aniline magnesium bromide, 0.5M solution in THF, AcroSeal™
4-(N,N-Dimethyl)aniline magnesium bromide, 0.5M solution in THF, AcroSeal™
4-(N,N-Dimethyl)aniline magnesium bromide, 0.5M solution in THF, AcroSeal™
Acros Organics

4-(N,N-Dimethyl)aniline magnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 7353-91-5 | C8H10BrMgN | 224.38 g/mol
Have Questions?
Catalog NumberQuantity
45061050050 mL
Catalog number 450610500
HSN 98020000
Price (INR)
22,007.00
Each
Quantity:
50 mL
Request bulk or custom format
Price (INR)
22,007.00
Each
Specifications
Chemical Name or Material4-(N, N-Dimethyl)aniline magnesium bromide
PackagingAcroSeal™ Glass bottle
CAS7353-91-5
Density0.9510g/mL
Flash Point−17°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only