TOPO™ TA Cloning™ Kit, Dual Promoter, without competent cells
Invitrogen™

TOPO™ TA Cloning™ Kit, Dual Promoter, without competent cells

TOPO™ TA Cloning™ Dual Promoter kits are for fast, efficient cloning and subsequent in vitro transcription. These kits include theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45064025 Reactions
45264010 Reactions
Catalog number 450640
Price (USD)
449.65
Online exclusive
484.00
Save 34.35
Each
Quantity:
25 Reactions
Price (USD)
449.65
Online exclusive
484.00
Save 34.35
Each
TOPO™ TA Cloning™ Dual Promoter kits are for fast, efficient cloning and subsequent in vitro transcription. These kits include the pCR™II-TOPO™ TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO™ cloning is the most reliable method, featuring a 3-step protocol and 5-minute cloning reaction, and yielding up to 95% recombinants.

Convenient features of this TOPO™ TA Cloning™ Dual Promoter kit include:
• 3´-T overhangs for direct ligation of Taq-amplified PCR products
• T7 and SP6 promoters for efficient in vitro transcription
• M13 forward and reverse primer sites for sequencing
• 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
• Kanamycin and ampicillin resistance for your choice of selection in E. coli
• Easy blue/white colony screening for selection of recombinants
• No competent cells included for flexibility and cost-savings

Kit Options: TOPO™ TA Cloning™ Dual Promoter Kits
TOPO™ TA Cloning™ Dual Promoter kits can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:
• General cloning: TOP10 cells (Cat. Nos. K4600-J10, K4600-01, K4600-40)
• High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. Nos. K4660-01, K4660-40)
• General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. Nos. K4620-01, K4620-40)
• Fast growth: Mach1™ -T1R chemically competent E. coli (Cat. No. K4610-20)
• Repressor/induction needs: TOP10F’ cells (Cat. Nos. K4650-01, K4650-40)
• Provide your own cells (Cat. No. 450640)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR), Kanamycin (KanR)
Cloning MethodTOPO™-TA
Product LineTOPO
Product TypeCloning Kit
PromoterT7, SP6
Quantity25 Reactions
Shipping ConditionDry Ice
VectorTOPO-TA Vectors
FormatKit
Unit SizeEach
Contents & Storage
Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Citations & References (1)

Citations & References
Abstract
Characterization of the coronavirus mouse hepatitis virus strain A59 small membrane protein E.
Authors:Raamsman MJ, Locker JK, de Hooge A, de Vries AA, Griffiths G, Vennema H, Rottier PJ
Journal:J Virol
PubMed ID:10666264
The small envelope (E) protein has recently been shown to play an essential role in the assembly of coronaviruses. Expression studies revealed that for formation of the viral envelope, actually only the E protein and the membrane (M) protein are required. Since little is known about this generally low-abundance virion ... More