TOPO™ TA Cloning™ Kit for Subcloning, without competent cells
Invitrogen™

TOPO™ TA Cloning™ Kit for Subcloning, without competent cells

TOPO™ TA Cloning™ Kits for Subcloning provide a highly efficient, 5-minute, one-step cloning strategy ('TOPO™ cloning') for the direct insertionRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45064125 Reactions
45164110 Reactions
Catalog number 450641
Price (USD)
450.65
Online exclusive
485.00
Save 34.35
Each
Quantity:
25 Reactions
Price (USD)
450.65
Online exclusive
485.00
Save 34.35
Each
TOPO™ TA Cloning™ Kits for Subcloning provide a highly efficient, 5-minute, one-step cloning strategy ('TOPO™ cloning') for the direct insertion of Taq polymerase–amplified PCR products into a plasmid vector for subcloning. Each kit uses the pCR™ 2.1-TOPO™ TA vector with convenient restriction sites for subcloning. The cloning kits are available with a variety of competent cells, or without competent cells, depending upon your needs and budget. Features of the TOPO™ TA Cloning™ Kits for Subcloning:

• Fast and easy—go from PCR to clones in just 3 steps and in as little as 5 minutes hands-on time
• Efficient—obtain up to 95% clones with correct insert
• Proven—reliable performance for over a decade with over 4,000 citations
• Simple—no ligase, post-PCR procedures, or PCR primers containing specific sequences are required

TOPO™ TA Cloning™ Kits for Subcloning—overview

• Vector: pCR™ 2.1-TOPO™ TA vector—subcloning vector with 15 convenient restriction sites flanking your insert for easy, directional subcloning
• Cloning method: TOPO™ TA Cloning™ —Topoisomerase I–based, 5-minute ligation of PCR products with 3′-A overhangs (Taq polymerase amplified) to TOPO™ vector with T overhangs
• Competent cells: various options—choose from kits with either general, high-efficiency, bacteriophage T1–resistant, fast-growing competent cells, or use your own

pCR™ 2.1-TOPO™ TA vector—cloning and subcloning simplicity
The pCR™ 2.1-TOPO™ TA vector is linearized with 3'-thymidine (T) overhangs for direct ligation of Taq polymerase–amplified PCR products (TA cloning™) and is 'activated' with covalently bound Topoisomerase I. There is no need to add ligase and cloning is complete in 5 minutes. EcoRI sites flanking the PCR product insertion site allow for easy excision of inserts or use any combination of 15 convenient restriction sites flanking your PCR insert for easy, directional subcloning.

pCR™ 2.1-TOPO™ TA clone selection and manipulation
The pCR™ 2.1-TOPO™ TA vector contains both ampicillin and kanamycin resistance markers and the LacZα gene for blue/white screening.

Simplified TOPO™ -based cloning
Using TOPO™ cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform E. coli competent cells.

Efficient cloning
With up to 95% of clones carrying the desired insert, you can screen less clones, saving time and money. The pCR™ 2.1-TOPO™ TA vector used in this kit comes with 3'-T overhangs for efficient ligation of Taq polymerase–amplified PCR products, which contain 3'-A overhangs.

The standard in cloning
When it comes to cloning, TOPO™ cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO™ cloning has been applied to many different vectors for a wide array of applications.

TOPO™ TA Cloning™ Kits for Subcloning—kit options
The TOPO™ TA Cloning™ Kit for Sequencing can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

• General cloning: TOP10 cells (Cat. No. K4500-01, K4500-40)
• High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K4560-01, K4560-40)
• General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. No. K4520-01, K4520-40)
• Fast growth: Mach1™ -T1R Chemically Competent E. Coli (Cat. No. K4510-20)
• Provide your own: for flexibility and to save money (Cat. No. 450641)

We also offer two versions of the kit that include a PureLink™ Quick Plasmid Miniprep Kit (Cat. No. K4500-02 and K4510-02) for use in isolation of clean, sequencing-ready, recombinant plasmid.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR), Kanamycin (KanR)
Bacterial or Yeast StrainNot Included
Cloning MethodTOPO™-TA
Product LineTOPO
Product TypeCloning Kit
Quantity25 Reactions
Shipping ConditionDry Ice
VectorpCR, TOPO-TA Cloning Vectors
FormatKit
Unit SizeEach
Contents & Storage
Box 1:
• Topoisomerase I-activated pCR™2.1-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Frequently asked questions (FAQs)

Can I store my competent E. coli in liquid nitrogen?

We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.

How should I store my competent E. coli?

We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.

What is the difference between the pCR2.1-TOPO and pCR4-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCR4-TOPO vector has sequencing primer sites located as close as 33 base pairs from the PCR product insertion site. This minimizes the amount of vector DNA sequence that needs to be read before reaching the sequence of the insert, making the pCR4-TOPO vector very useful for sequencing applications.

What is the difference between the pCR2.1-TOPO and pCRII-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCRII-TOPO vector is a dual promoter vector, containing the SP6 and T7 promoters for in vitro transcription/sequencing, whereas the pCR2.1-TOPO vector contains only the T7 promoter for in vitro transcription/sequencing. Both vectors contain the M13 Forward and Reverse primer sites for sequencing or PCR screening.

Your TOPO cloning kits contain a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

Citations & References (6)

Citations & References
Abstract
Efficacy, safety, and pharmacokinetics of imatinib dose escalation to 800 mg/day in patients with advanced gastrointestinal stromal tumors.
Authors:Yoo C, Ryu MH, Ryoo BY, Beck MY, Kang YK
Journal:Invest New Drugs
PubMed ID:23591629
'Imatinib dose escalation has been suggested as an effective therapy for advanced gastrointestinal stromal tumors (GIST) after progression on the standard dose. We evaluated the efficacy, tolerability, and pharmacokinetics of imatinib dose escalation. Eighty-four patients with GIST who received imatinib 800 mg/day as second-line therapy were reviewed. In 66 patients, imatinib ... More
Use of cell-SELEX to generate DNA aptamers as molecular probes of HPV-associated cervical cancer cells.
Authors:Graham JC, Zarbl H
Journal:PLoS One
PubMed ID:22536456
'Disease-specific biomarkers are an important tool for the timely and effective management of pathological conditions, including determination of susceptibility, diagnosis, and monitoring efficacy of preventive or therapeutic strategies. Aptamers, comprising single-stranded or double-stranded DNA or RNA, can serve as biomarkers of disease or biological states. Aptamers can bind to specific ... More
A streamlined method for detecting structural variants in cancer genomes by short read paired-end sequencing.
Authors:Mijuškovic M, Brown SM, Tang Z, Lindsay CR, Efstathiadis E, Deriano L, Roth DB
Journal:PLoS One
PubMed ID:23144753
Defining the architecture of a specific cancer genome, including its structural variants, is essential for understanding tumor biology, mechanisms of oncogenesis, and for designing effective personalized therapies. Short read paired-end sequencing is currently the most sensitive method for detecting somatic mutations that arise during tumor development. However, mapping structural variants ... More
Two CRISPR-Cas systems in Methanosarcina mazei strain Gö1 display common processing features despite belonging to different types I and III.
Authors:Nickel L, Weidenbach K, Jäger D, Backofen R, Lange SJ, Heidrich N, Schmitz RA
Journal:RNA Biol
PubMed ID:23619576
The clustered regularly interspaced short palindromic repeats (CRISPR) system represents a highly adaptive and heritable defense system against foreign nucleic acids in bacteria and archaea. We analyzed the two CRISPR-Cas systems in Methanosarcina mazei strain Gö1. Although belonging to different subtypes (I-B and III-B), the leaders and repeats of both ... More
Camphor pathway redux: functional recombinant expression of 2,5- and 3,6-diketocamphane monooxygenases of Pseudomonas putida ATCC 17453 with their cognate flavin reductase catalyzing Baeyer-Villiger reactions.
Authors:Iwaki H, Grosse S, Bergeron H, Leisch H, Morley K, Hasegawa Y, Lau PC
Journal:Appl Environ Microbiol
PubMed ID:23524667
Whereas the biochemical properties of the monooxygenase components that catalyze the oxidation of 2,5-diketocamphane and 3,6-diketocamphane (2,5-DKCMO and 3,6-DKCMO, respectively) in the initial catabolic steps of (+) and (-) isomeric forms of camphor (CAM) metabolism in Pseudomonas putida ATCC 17453 are relatively well characterized, the actual identity of the flavin ... More