Ammonium bromide, 98%, extra pure
Ammonium bromide, 98%, extra pure
Ammonium bromide, 98%, extra pure
Thermo Scientific Chemicals

Ammonium bromide, 98%, extra pure

CAS: 12124-97-9 | BrH4N | 97.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 450650010
Price (USD)
67.60
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
67.60
Each
Chemical Identifiers
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Sulfated ash=<0.1 %
Appearance (Color)White
Titration Argentometric>=97.5 %
Insoluble matter=<0.01 % (in water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only