Informational:Try chem dex – the new web app for synthetic chemists
Thermo Scientific Chemicals

Lithium triethylborohydride, 1.7M solution in THF, Thermo Scientific Chemicals

Catalog number: 450691000
100 mL, Each
Thermo Scientific Chemicals

Lithium triethylborohydride, 1.7M solution in THF, Thermo Scientific Chemicals

Catalog number: 450691000
100 mL, Each
Quantity

Chemical Identifiers

CAS
22560-16-3
IUPAC Name
lithium(1+) triethylboranuide
Molecular Formula
C6H16BLi
InChI Key
WCJAYABJWDIZAJ-UHFFFAOYSA-N
SMILES
[Li+].CC[BH-](CC)CC
Appearance (Color)
Hazy colorless to light grey
Strength
1.6 to 1.8 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text